Learn More
Ansell AlphaTec™ 5000
Product Code. 17190885
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
O50T103
O50T111
O50T122
O50T125B
O50T151
O50T151B
Unit Size:
Pack of 10
Pack of 8
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Style unitSize
17190885 Large O50T151B Pack of 8
17190855 Small O50T103 Pack of 10
17100865 Medium O50T103 Pack of 10
17110865 Large O50T103 Pack of 10
17120865 X-Large O50T103 Pack of 10
17130865 2X-Large O50T103 Pack of 10
17140865 3X-Large O50T103 Pack of 10
17150865 Small O50T111 Pack of 10
17120875 Small O50T122 Pack of 8
17130875 Medium O50T122 Pack of 8
17140875 Large O50T122 Pack of 8
17150875 X-Large O50T122 Pack of 8
17160875 2X-Large O50T122 Pack of 8
17170875 3X-Large O50T122 Pack of 8
17180875 Small O50T125B Pack of 8
17190875 Medium O50T125B Pack of 8
17100885 Large O50T125B Pack of 8
17110885 X-Large O50T125B Pack of 8
17120885 2X-Large O50T125B Pack of 8
17130885 3X-Large O50T125B Pack of 8
17140885 Small O50T151 Pack of 8
17160885 Medium O50T151 Pack of 8
17180885 Large O50T151 Pack of 8
17100895 X-Large O50T151 Pack of 8
17120895 2X-Large O50T151 Pack of 8
17140895 3X-Large O50T151 Pack of 8
17150885 Small O50T151B Pack of 8
17170885 Medium O50T151B Pack of 8
17110895 X-Large O50T151B Pack of 8
17130895 2X-Large O50T151B Pack of 8
17150895 3X-Large O50T151B Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
Product Code. 17190885 Supplier Ansell Supplier No. OR50T0015104G02
Only null left
Add to basket
Add to basket
This highly visible innovative material is strong, durable and suitable for workers in extremely hazardous areas, including HAZMAT response teams.
  • Outstanding permeation performance to numerous organic and inorganic chemicals, biological hazards and chemical warfare agents
  • Exceptional barrier protection, providing >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
  • Designed to protect, rear entry suit with neoprene rubber face seal for a close fit to full face respirators
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Designed to protect, typical innovative features include dual zip systems, double cuffs with knitted inner cuffs
  • Multi-layer barrier material which is lightweight, yet strong and durable
  • Anti-static
TRUSTED_SUSTAINABILITY
Specifications
Size Large
Material Multi-layer Barrier Laminate Fabric
Color Orange
Seam Type Welded, Taped
Certifications/Compliance (Europe) EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003, NFPA 1992
Includes Neoprene rubber face seal
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes
Weight (Metric) 132 gsm
Style O50T151B