missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- The AlphaTec™ 3000 Ultrasonically Welded & Taped- Model 103 protective suit features multi-layered fabric that ensures an effective barrier against numerous inorganic chemicals and biohazards
- This protective barrier is further reinforced by its double cuffs, dual-zip systems and double zip enclosure, which extends wear life
- Highly visible bright yellow color, this protective outer garment remains particularly noticeable in low-lit environments, enhancing wearer and workplace safety
- In addition, it complies with EN 1149-5 antistatic standards
Specifications
Specifications
| Product Type | Chemical Protective Clothing |
| Size | Small |
| Gender | Unisex |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Yellow |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Double, Elastic |
| Neck Style | Collared |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?