Learn More
Ansell AlphaTec™ 3000 Ultrasonically Welded Model 122
Product Code. p-200007430 Shop All Ansell Products
Click to view available options
Size:
4X-Large
5X-Large
Unit Size:
Each
Product Code. 17919150
Brand: Ansell YE30W0012208
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
AlphaTec™ 3000 Ultrasonically Welded Model 122 features a multi-layer fabric barrier for protection from various chemicals.
  • Specialized protection: Made with durable multi-layer nonwoven laminate, the AlphaTec™ 3000 Ultrasonically Welded, Model 122 protects against various inorganic chemicals and biological hazards, while ensuring lightweight comfort
  • Enhanced features: This chemical-protective suit features dual-zip systems, double cuffs and attached barrier gloves, enhancing its protective capabilities, as well as a two-piece hood and an elasticated waist, sleeves and over-cuffs
  • Improved visibility: Thanks to its high-visibility yellow exterior, this protective suit ensures greater wearer and workplace safety
  • Assured antistatic capabilities: This full-body protection suit is also tested against EN 1149-5 antistatic standards
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Protective Clothing
Size 4X-Large
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Reusable
Waist Style Elastic
Ankle Style Attached Socks, Boot Overflaps
Cuff Style Elastic
Neck Style Hooded
Hood Type 2-panel, Elastic
Closure Type Double Zipper
Seam Type Welded
Sterility Non-sterile
Protection Class Type 3-B, Type 4-B, Type 5-B
Resistance Liquid, Chemicals, Particle, Anti-static, Bloodborne Pathogens
Construction Coverall with 2-piece Hood, Double Zip Closure, Elasticated Hood, Waist and Sleeve Over Cuffs, and Integrated Socks with Boot Overflap
Certifications/Compliance (Europe) CE Number, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2018, EN ISO 13982-1:2004 + A1:2010, EN 14126:2003
Latex Free No
No. of Pockets None
For Use With (Application) Transferring liquids and solids between vessels and tanks and process equipment, Unexpected leakages spills or other releases, Opening and draining pumps valves or lines, Loading and unloading trucks and vehicles, Filling, blending and charging of raw materials, Product inspection, Mixing or loading chemicals, Cleaning of plant and machines
Style 3000 122