Learn More
Ansell AlphaTec 3000 Coverall, Hood
Product Code. 16855020
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 20
Pack of 25
Product Code. 16855020
Brand: Ansell YE30W0012106
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical protective suits with a durable, comfortable multi-layer chemical barrier, and Type 3/4/5 protection.
  • Designed to protect, typical innovative features include dual zip systems and double cuffs
  • Lightweight and durable
  • Highly visible bright yellow color for improved worker safety 
  • Anti-static
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Seam Type Welded
Certifications/Compliance (Europe) EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003
Model Alphatec Model 121
For Use With (Application) Transferring Liquids and Solids Between Vessels and Tanks and Process Equipment, Unexpected Leakages Spills or Other Releases, Opening and Draining Pumps Valves or Lines, Loading and Unloading Trucks and Vehicles, Filling, Blending and Charging of Raw Materials, Product Inspection, Mixing or Loading Chemicals, Cleaning of Plant and Machines
Weight (Metric) 80 gsm
Style Y30W121