Learn More
Ansell AlphaTec™ 2300 STANDARD Bound
Product Code. 16895060
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Style:
Y23B103
Y23B111
Unit Size:
Pack of 40
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Style unitSize
16895060 Large Y23B111 Pack of 40
16895050 Small Y23B103 Pack of 40
16805060 Medium Y23B103 Pack of 40
16815060 Large Y23B103 Pack of 40
16825060 X-Large Y23B103 Pack of 40
16835060 2X-Large Y23B103 Pack of 40
16845060 3X-Large Y23B103 Pack of 40
16855060 4X-Large Y23B103 Pack of 40
16865060 5X-Large Y23B103 Pack of 40
16875060 Small Y23B111 Pack of 40
16885060 Medium Y23B111 Pack of 40
16805070 X-Large Y23B111 Pack of 40
16815070 2X-Large Y23B111 Pack of 40
16825070 3X-Large Y23B111 Pack of 40
16835070 4X-Large Y23B111 Pack of 40
16845070 5X-Large Y23B111 Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 16895060 Supplier Ansell Supplier No. YY23B0011104
Add to basket
Add to basket
Chemical protective suits with a good protective barrier to numerous inorganic liquid chemicals including acids and bases.
  • Lightweight, yet relatively strong and durable
  • Designed to protect, typical coverall features include respirator fit hood and a zip flap with self-adhesive tape closure
  • Anti-static
TRUSTED_SUSTAINABILITY
Specifications
Size Large
Material Polyethylene on Bicomponent Polypropylene, Polyethylene Non-Woven
Color Yellow
Seam Type Bound
Certifications/Compliance (Europe) EN 374:2003, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003
For Use With (Application) Environmental Clean-up and Remediation, Assembly and Disassembly of Large Pipes, Transferring Liquids and Solids, Assembling and Mounting Components
Weight (Metric) 65 gsm
Style Y23B111