Learn More
Ansell™ AlphaTec™ 2000 Ts PLUS Stitched and Taped
Product Code. 16854820
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 30
Product Code. 16854820
Brand: Ansell™ WH20T0015603
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
AlphaTec™ 2000 Ts PLUS Stitched and taped is designed to allow water vapor (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals and filter 100% of particulates down to 0.01 microns in size.
Designed to allow perspiration to escape from the suit and withstand saturation of liquid chemicals.
  • Filters 100% of particulates down to 0.01 microns in size
  • Proven protective barrier to low concentration liquid chemicals, liquid, and particulate biological hazards
  • Comfortable to wear featuring moisture vapor permeable fabric to reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Anti-static, tested according to EN 1149-5
  • Ultra Low Linting, reduced risk of contamination in critical areas
  • Silicone free, critical in spray painting applications
  • Tunnelled elasticated hood, wrists, and ankles to minimize the risk of linting and cross contamination
TRUSTED_SUSTAINABILITY
Specifications
Product Type Controlled Environments ESD Coveralls
Size Medium
Gender Unisex
Material Microporous Polyethylene Laminate (Non-woven)
Color White
Disposable Reusable
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Stitched, Taped
Sterility Non-sterile
Protection Class CE CATEGORY III
Resistance Chemicals
Collar Type Hood
Certifications/Compliance CE CATEGORY III (Type 4-B, Type 5-B, Type 6-B, EN 1073-2, EN 14126, DIN 32781, EN 1149-5)
No. of Pockets None