missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ AlphaTec™ 2000 Ts PLUS SOCO Stitched and Taped
442.09 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
Designed to allow perspiration to escape from the suit and withstand saturation of liquid chemicals
- Filter 100% of particulates down to 0.01 microns in size.
- Proven protective barrier to low concentration liquid chemicals, liquid, and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric to reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Anti-static, tested according to EN 1149-5
- Ultra Low Linting, reduces rinks of crime scene contamination
- Silicone free, critical in spray painting applications
- Tunnelled elasticated hood, wrists, and ankles to minimize the risk of linting and cross contamination
Specifications
Product Type | Controlled Environments ESD Coveralls |
Size | Small |
Gender | Unisex |
Material | Microporous Non-woven Polyethylene Laminate |
Color | White |
Disposable | Reusable |
Ankle Style | Elastic |
Cuff Style | Elastic, Finger Loops |
Neck Style | Hooded |
Closure Type | Zipper |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'