Learn More
Ansell AlphaTec™ 2000 STANDARD Bound
Product Code. 17121015
Change view
Click to view available options
Size:
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
8X-Large
Large
Medium
One Size
Small
X-Large
X-Small
Style:
W20B102
W20B103
W20B111
W20B113
W20B122
W20B162
Unit Size:
Pack of 30
Pack of 35
Pack of 40
Color:
White
White, Green
33 product options available for selection
Product selection table with 33 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Style unitSize Color
17121015 3X-Large W20B122 Pack of 30 -
16864750 Small W20B102 Pack of 40 White
16874750 Medium W20B102 Pack of 40 White
16884750 Large W20B102 Pack of 40 -
16894750 X-Large W20B102 Pack of 40 White
16804760 2X-Large W20B102 Pack of 40 White
16814760 3X-Large W20B102 Pack of 40 White
16824760 4X-Large W20B102 Pack of 40 White
16834760 5X-Large W20B102 Pack of 40 White
16804770 Small W20B113 Pack of 35 White
16814770 Medium W20B113 Pack of 35 White
16824770 Large W20B113 Pack of 35 White
16834770 X-Large W20B113 Pack of 35 White
16844770 2X-Large W20B113 Pack of 35 White
16854770 3X-Large W20B113 Pack of 35 -
16864770 4X-Large W20B113 Pack of 35 White
16874770 5X-Large W20B113 Pack of 35 White
16251422 5X-Large W20B103 Pack of 40 White, Green
16844760 X-Small W20B111 Pack of 40 White, Green
16854760 4X-Large W20B111 Pack of 40 White, Green
16864760 5X-Large W20B111 Pack of 40 White, Green
16874760 6X-Large W20B111 Pack of 30 White, Green
16884760 7X-Large W20B111 Pack of 30 White, Green
16894760 8X-Large W20B111 Pack of 30 White, Green
16844780 Small W20B162 Pack of 40 White
17111015 X-Small W20B122 Pack of 30 White
16854780 Medium W20B162 Pack of 40 White
16864780 Large W20B162 Pack of 40 White
16874780 X-Large W20B162 Pack of 40 White
16884780 2X-Large W20B162 Pack of 40 White
16894780 3X-Large W20B162 Pack of 40 White
16814790 5X-Large W20B162 Pack of 40 White
16804790 One Size W20B162 Pack of 40 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 33 options available.
33 options
Product Code. 17121015 Supplier Ansell Supplier No. WH20B0012207
Only null left
Add to basket
Add to basket
Chemical protective suits that have a proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards.
  • Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Low linting to help reduce the risk of cross contamination in critical areas
  • Silicone free, critical in spray painting applications
TRUSTED_SUSTAINABILITY
Specifications
Size 3X-Large
Material Microporous Polyethylene Laminate (Non-woven)
Seam Type Bound
Certifications/Compliance (Europe) EN 374:2003, EN ISO 13982-1:2004 + A1:2010,EN 13034:2005 + A1:2009,EN 1073-2:2002,EN 1149-5:2018,EN 14126:2003,EN ISO 13688:2013,DIN 32781:2010
For Use With (Application) Clean Room Cleaning and Preparing, Body Painting Including Surface Inspection, Production Line Support and Maintenance, Cleaning of Plant and Machines, Maintenance of Plant and Machines, Crime Scene Investigation (CSI), Veterinary Services
Weight (Metric) 63 gsm
Style W20B122