missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 1800 Ts PLUS
Description
- 3 piece hood
- Elasticated hood, wrists, waist and ankles
- 2-way front zipper with resealable storm flap
- Finger Loops
- Chinstrap
Specifications
Specifications
| Size | 2X-Large |
| Material | Microporous Polyethylene Laminate (Non-woven) |
| Color | White |
| Seam Type | Stitched, Taped |
| Includes | Hood and Boots |
| For Use With (Application) | Clean Room Cleaning and Preparing, Body Painting Including Surface Inspection, Production Line Support & Maintenance, Cleaning of Plant and Machines, Maintenance of Plant and Machines, Fibre Glass Resin Applications, Handling of Powders |
| Weight (Metric) | 53 gsm |
| Style | W18T122 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?