missing translation for 'onlineSavingsMsg'
Learn More

(AlphaR,1S,3S,4R)-Alpha,4-Dimethyl-2-oxo-3-(3-oxobutyl)-cyclohexaneacetic Acid, TRC

Product Code. 30386721
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386721 5 mg 5mg
30380360 25 mg -
30374397 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386721 Supplier Toronto Research Chemicals Supplier No. TRCD4769555MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 173427-03-7
Molecular Formula C14H22O4
Molecular Weight (g/mol) 254.32
Synonym Dihydroartemisin Impurity F
SMILES C[C@H]([C@@H]1CC[C@@H](C)[C@H](CCC(=O)C)C1=O)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H22O4/c1-8-4-6-12(10(3)14(17)18)13(16)11(8)7-5-9(2)15/h8,10-12H,4-7H2,1-3H3,(H,17,18)/t8-,10-,11+,12+/m1/s1
Formula Weight 254.1518
Chemical Name or Material (alphaR,1S,3S,4R)-alpha,4-Dimethyl-2-oxo-3-(3-oxobutyl)-cyclohexaneacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.