CAS RN 173427-03-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(AlphaR,1S,3S,4R)-Alpha,4-Dimethyl-2-oxo-3-(3-oxobutyl)-cyclohexaneacetic Acid, TRC
CAS: 173427-03-7 Molecular Formula: C14H22O4 Molecular Weight (g/mol): 254.32 Synonym: Dihydroartemisin Impurity F SMILES: C[C@H]([C@@H]1CC[C@@H](C)[C@H](CCC(=O)C)C1=O)C(=O)O