missing translation for 'onlineSavingsMsg'
Learn More

Skytec™ Alphachem X150 Disposable Coverall

Product Code. 31702844
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 10

Product Code. 31702844

Brand: Skytec™ CDX1B/HX1500/YE2

Please to purchase this item. Need a web account? Register with us today!

X150 disposable garments offer dependable protection for a range of chemical-related tasks.

  • Provide increased wearer comfort and exceptional levels of safety and protection, with superior durability
  • Double sleeve system with inner thumb loop
  • Improved fit, enhanced freedom of movement with two-piece crotch pattern
  • A 3-piece hood ensures a comfortable fit
  • Double zip/grab tag, easier to hold when wearing gloves
  • Knee patches give extra reinforcement where it's needed most
  • Stitched and taped seams provide a strong and effective chemical barrier
  • Quick and easy donning/doffing
  • Elasticated ankles and wrists ensure a snug and secure fit

Specifications

Product Type Chemical-resistant Coveralls
Size Medium
Gender Unisex
Material Composite Fabric
Color Yellow
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Zipper
Seam Type Stitched, Taped
Sterility Non-sterile
Resistance Anti-static, Particle, Liquid, Chemicals, Viruses
Certifications/Compliance CE III or - EN 1073-2:2002 CLASS 1 or - EN 1149-5:2018 or - EN 13034:2005 + A1:2010 Type 6 or - EN 13982-1:2004 + A1:2010 Type 5 or - EN 14126:2003 + AC:2004 Type 3B,Type 4B,Type 5B,Type 6B or - EN 14605:2005 + A1:2009 Type 3,Type 4 or - EN 27065:2017 + A1:2019
For Use With (Application) Chemical Handling and Containment, Commercial Cleaning, Commercial Washdown
Weight (Metric) 500 g
Show More Show Less
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.