missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha smooth muscle actin specific Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | Alpha smooth muscle actin specific |
|---|---|
| Clone | 1E9A11 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235764
|
Proteintech
67735-1-IG-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30238047
|
Proteintech
67735-1-IG-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Alpha smooth muscle actin specific antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Alpha smooth muscle actin specific. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The Alpha smooth muscle actin specific antibody has been validated for the following applications: WB, IHC, IF/ICC, IF-P, FC (Intra), ELISA analysis.Specifications
| Alpha smooth muscle actin specific | |
| 500 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 59 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1E9A11 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Pig, Rabbit | |
| AAT6; ACTSA | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title