missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha smooth muscle actin specific Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67735-1-IG-20UL
This item is not returnable.
View return policy
Description
The Alpha smooth muscle actin specific antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Alpha smooth muscle actin specific. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The Alpha smooth muscle actin specific antibody has been validated for the following applications: WB, IHC, IF/ICC, IF-P, FC (Intra), ELISA analysis.Specifications
| Alpha smooth muscle actin specific | |
| Monoclonal | |
| 500 μg/mL | |
| AAT6; ACTSA | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat, Pig, Rabbit | |
| 42 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| 1E9A11 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 59 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction