missing translation for 'onlineSavingsMsg'
Learn More

alpha N Catenin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen alpha N Catenin
Concentration 0.33 mg/mL
Applications Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16832974
View Documents
Proteintech
14362-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16822974
View Documents
Proteintech
14362-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The catenins (alpha, beta, and gamma) are cytoplasmic proteins found in varying abundance in many developing and adult tissues. Catenins bind directly or indirectly to the conserved cytoplasmic tail domain of the cell adhesion cadherins. The association of catenins to cadherins produces a complex, which is linked to the actin filament network. Catenins/cadherin complexes play an important role in mediating cell adhesion, transduction of cell-cell contact positional signals to the cell interior, and may play a crucial role in cell differentiation. Alpha-Catenin (CAP102, 102 kDa), originally described as an E-cadherin associated protein, has been shown to associate with other members of the cadherin family members, N-cadherin and P-cadherin.
Specifications

Specifications

alpha N Catenin
Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Mouse, Human, Rat
P26232, Q61301
12386, 1496, 297357
alpha N Catenin Fusion Protein Ag5747
Primary
-20°C
CTNNA2
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
alpha 2 Catenin, Alpha N catenin, Catenin alpha 2, CTNNA2
Ctnna2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.