missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha N Catenin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14362-1-AP-20UL
This item is not returnable.
View return policy
Description
The catenins (alpha, beta, and gamma) are cytoplasmic proteins found in varying abundance in many developing and adult tissues. Catenins bind directly or indirectly to the conserved cytoplasmic tail domain of the cell adhesion cadherins. The association of catenins to cadherins produces a complex, which is linked to the actin filament network. Catenins/cadherin complexes play an important role in mediating cell adhesion, transduction of cell-cell contact positional signals to the cell interior, and may play a crucial role in cell differentiation. Alpha-Catenin (CAP102, 102 kDa), originally described as an E-cadherin associated protein, has been shown to associate with other members of the cadherin family members, N-cadherin and P-cadherin.Specifications
| alpha N Catenin | |
| Polyclonal | |
| Unconjugated | |
| CTNNA2 | |
| alpha 2 Catenin, Alpha N catenin, Catenin alpha 2, CTNNA2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12386, 1496, 297357 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P26232, Q61301 | |
| Ctnna2 | |
| alpha N Catenin Fusion Protein Ag5747 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction