missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylbenzylzinc chloride, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15425335
Change view
Click to view available options
Quantity:
50 mL
Packaging:
ChemSeal™ bottles
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15425335 50 mL ChemSeal™ bottles 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15425335 Supplier Thermo Scientific Alfa Aesar Supplier No. H58185.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It helps in the synyhesis of of heterotactic polylactic acid from rac-lactide.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases and ox
TRUSTED_SUSTAINABILITY

Specifications

CAS 117892-77-0
Color Black to Yellow
Density 0.965 g/mL
Packaging ChemSeal™ bottles
Molecular Formula C8H9BrZn
MDL Number MFCD01311436
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym a-methylbenzylzinc bromide, bromo 1-phenylethyl zinc, alpha-methylbenzyl zinc bromide, alpha-methylbenzylzinc chloride, a-methylbenzylzinc bromide 0.5 m in tetrahydrofuran, alpha-methylbenzylzinc bromide solution, 0.5 m in thf
Solubility Information Reacts with water.
InChI Key LCPVHNNFHPWFCI-UHFFFAOYSA-M
SMILES C[CH-]C1=CC=CC=C1.[Zn+]Br
IUPAC Name bromozinc(1+);ethylbenzene
Molecular Weight (g/mol) 250.444
PubChem CID 4524692
Formula Weight 206
Physical Form Liquid
Chemical Name or Material alpha-Methylbenzylzinc chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.