missing translation for 'onlineSavingsMsg'
Learn More

alpha-Fluoro-beta-alanine, TRC

Product Code. 30003765
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003765 250 mg 250mg
30004803 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003765 Supplier Toronto Research Chemicals Supplier No. TRCF587350250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3821-81-6
Molecular Formula C3 H6 F N O2
Molecular Weight (g/mol) 107.08
Synonym Propanoic acid, 3-amino-2-fluoro-, β-Alanine, 2-fluoro- (6CI,7CI,8CI), 3-Amino-2-fluoropropanoic acid, 2-Fluoro-β-alanine, Fluoro-β-alanine, α-Fluoro-β-alanine
IUPAC Name 3-amino-2-fluoropropanoic acid
SMILES NCC(F)C(=O)O

Specifications

InChI Formula InChI=1S/C3H6FNO2/c4-2(1-5)3(6)7/h2H,1,5H2,(H,6,7)
Quantity 250 mg
Formula Weight 107.0383
Shipping Condition Room Temperature
Chemical Name or Material a-Fluoro-beta-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.