CAS RN 3821-81-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
alpha-Fluoro-beta-alanine, TRC
CAS: 3821-81-6 Molecular Formula: C3 H6 F N O2 Molecular Weight (g/mol): 107.08 Synonym: Propanoic acid, 3-amino-2-fluoro-,β-Alanine, 2-fluoro- (6CI,7CI,8CI),3-Amino-2-fluoropropanoic acid,2-Fluoro-β-alanine,Fluoro-β-alanine,α-Fluoro-β-alanine IUPAC Name: 3-amino-2-fluoropropanoic acid SMILES: NCC(F)C(=O)O