missing translation for 'onlineSavingsMsg'
Learn More

alpha-Ethyl-3-nitrocinnamic acid, 99%, Thermo Scientific™

Product Code. 11388045
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388045 25 g 25g
11378045 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388045 Supplier Thermo Scientific Alfa Aesar Supplier No. L08221.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124525-55-9
Molecular Formula C11H11NO4
Molecular Weight (g/mol) 221.212
MDL Number MFCD00134948
InChI Key QMUHBYNLXQHZNR-RMKNXTFCSA-N
Synonym e-2-3-nitrobenzylidene butanoic acid, alpha-ethyl-3-nitrocinnamic acid, 2e-2-3-nitrophenyl methylidene butanoic acid, 2-3-nitrobenzylidene butanoic acid, .alpha.-ethyl-3-nitrocinnamic acid, ^a-ethyl-3-nitrocinnamic acid, cinnamic acid, .alpha.-ethyl-m-nitro, butanoic acid, 2-3-nitrophenyl methylene, 2e-2-ethyl-3-3-nitrophenyl prop-2-enoic acid
PubChem CID 6059692
IUPAC Name (2E)-2-[(3-nitrophenyl)methylidene]butanoic acid
SMILES CCC(=CC1=CC(=CC=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 138°C to 142°C
Quantity 25 g
Beilstein 2618101
Formula Weight 221.21
Percent Purity 99%
Chemical Name or Material alpha-Ethyl-3-nitrocinnamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.