missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Adaptin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | alpha Adaptin |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
alpha Adaptin Polyclonal specifically detects alpha Adaptin in Human samples. It is validated for Western Blot.Specifications
| alpha Adaptin | |
| Polyclonal | |
| Rabbit | |
| 100 kDa coated vesicle protein A, Adapter-related protein complex 2 alpha-1 subunit, Adaptor protein complex AP-2 subunit alpha-1, adaptor-related protein complex 2, alpha 1 subunit, ADTAAAP2-ALPHA, alpha1-adaptin, Alpha-adaptin A, AP-2 complex subunit alpha-1, CLAPA1adaptin, alpha A, Clathrin assembly protein complex 2 alpha-A large chain, clathrin-associated/assembly/adaptor protein, large, alpha 1, Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit | |
| AP2A1 | |
| IgG | |
| This product is specific to Subunit or Isoform: alpha-1. |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 160 | |
| Synthetic peptides corresponding to AP2A1 (adaptor-related protein complex 2, alpha 1 subunit) The peptide sequence was selected from the C terminal of AP2A1. Peptide sequence ILAKLKRKKGPGAGSALDDGRRDPSSNDINGGMEPTPSTVSTPSPSADLL. | |
| Primary | |
| 105 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title