missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Adaptin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-68949
This item is not returnable.
View return policy
Description
alpha Adaptin Polyclonal specifically detects alpha Adaptin in Human samples. It is validated for Western Blot.
Specifications
| alpha Adaptin | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| AP2A1 | |
| Synthetic peptides corresponding to AP2A1 (adaptor-related protein complex 2, alpha 1 subunit) The peptide sequence was selected from the C terminal of AP2A1. Peptide sequence ILAKLKRKKGPGAGSALDDGRRDPSSNDINGGMEPTPSTVSTPSPSADLL. | |
| Affinity purified | |
| RUO | |
| 160 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| 100 kDa coated vesicle protein A, Adapter-related protein complex 2 alpha-1 subunit, Adaptor protein complex AP-2 subunit alpha-1, adaptor-related protein complex 2, alpha 1 subunit, ADTAAAP2-ALPHA, alpha1-adaptin, Alpha-adaptin A, AP-2 complex subunit alpha-1, CLAPA1adaptin, alpha A, Clathrin assembly protein complex 2 alpha-A large chain, clathrin-associated/assembly/adaptor protein, large, alpha 1, Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit | |
| Rabbit | |
| 105 kDa | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: alpha-1. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction