Learn More
Alpha Actinin Mouse, anti-Human, CoraLite Plus 488, Clone: 1E2E1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | Alpha Actinin |
|---|---|
| Clone | 1E2E1 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18044265
|
Proteintech
CL488-66895-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18007905
|
Proteintech
CL488-66895-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Alpha Actinin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Alpha Actinin. This antibody recognizes human, rat, mouse antigen. The Alpha Actinin antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| Alpha Actinin | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 87 | |
| IgG2b | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1E2E1 | |
| Western Blot | |
| Liquid | |
| Human, Rat, Mouse | |
| actinin, alpha 1, ACTN1, Alpha Actinin, Alpha actinin 1, F actin cross linking protein, FLJ40884, FLJ54432, Non muscle alpha actinin 1 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.