Learn More
Alpha Actinin Mouse, anti-Human, CoraLite Plus 488, Clone: 1E2E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66895-20UL
Description
The Alpha Actinin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Alpha Actinin. This antibody recognizes human, rat, mouse antigen. The Alpha Actinin antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| Alpha Actinin | |
| 1E2E1 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse | |
| 155 kDa | |
| IgG2b |
| Western Blot | |
| 1000 μg/mL | |
| actinin, alpha 1, ACTN1, Alpha Actinin, Alpha actinin 1, F actin cross linking protein, FLJ40884, FLJ54432, Non muscle alpha actinin 1 | |
| 20 μL | |
| 87 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |