missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Actinin 4 Antibody (7H6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£159.00
Specifications
| Antigen | Alpha Actinin 4 |
|---|---|
| Clone | 7H6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 μg/mL, Flow Cytometry 1 μg 106 cells, ELISA 1:100 - 1:2000, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin 4 μg/mL, Immunofluorescence 4 μg/mL |
| Applications | Western Blot |
Description
Alpha Actinin 4 Monoclonal specifically detects Alpha Actinin 4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| Alpha Actinin 4 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| actinin alpha4 isoform, actinin, alpha 4, ACTININ-4, alpha-actinin-4, DKFZp686K23158, F-actin cross-linking protein, focal segmental glomerulosclerosis 1, FSGS, FSGS1, Non-muscle alpha-actinin 4 | |
| ACTN4 | |
| IgG | |
| Protein G purified | |
| alpha Actinin 4 (7H6) |
| 7H6 | |
| Western Blot 2 μg/mL, Flow Cytometry 1 μg 106 cells, ELISA 1:100 - 1:2000, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin 4 μg/mL, Immunofluorescence 4 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| O43707 | |
| 81 | |
| Synthetic peptide corresponding to residues A(884) P Y Q G P D A V P G A L D(897) of human alpha Actinin 4. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title