missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Actinin 4 Antibody (7H6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-74352-0.025mg
This item is not returnable.
View return policy
Description
Alpha Actinin 4 Monoclonal specifically detects Alpha Actinin 4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Alpha Actinin 4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 2 μg/mL, Flow Cytometry 1 μg 106 cells, ELISA 1:100 - 1:2000, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin 4 μg/mL, Immunofluorescence 4 μg/mL | |
| actinin alpha4 isoform, actinin, alpha 4, ACTININ-4, alpha-actinin-4, DKFZp686K23158, F-actin cross-linking protein, focal segmental glomerulosclerosis 1, FSGS, FSGS1, Non-muscle alpha-actinin 4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 81 | |
| Human | |
| Purified |
| Western Blot | |
| 7H6 | |
| Unconjugated | |
| O43707 | |
| ACTN4 | |
| Synthetic peptide corresponding to residues A(884) P Y Q G P D A V P G A L D(897) of human alpha Actinin 4. | |
| 0.025 mg | |
| Primary | |
| alpha Actinin 4 (7H6) | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction