missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ alpha Actinin 3 Recombinant Rabbit Monoclonal Antibody (23GB3625), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£162.00 - £387.00

Specifications

Antigen alpha Actinin 3
Clone 23GB3625
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30089943
GREENER_CHOICE
View Documents
Invitrogen™
MA552586
100 μL
£387.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30087710
View Documents
Invitrogen™
MA552587
20 μL
£162.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

alpha Actinin 3 Recombinant Monoclonal antibody specifically detects alpha Actinin 3 in Human, Rat samples. It is validated for Immunocytochemistry, Western Blot
Specifications

Specifications

alpha Actinin 3
Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Rat
Q08043
171009, 89
IgG
Affinity Chromatography
Antibody
23GB3625
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
actinin alpha 3; actinin alpha 3 (gene/pseudogene); Actinin alpha3; actinin, alpha 3; Actinin-alpha3; ACTN3; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 3; alpha-Actinin3; Alpha-actinin-3; alphaActinin-3; F-actin cross-linking protein
A synthetic peptide of human ACTN3 (760-800AA).
Primary
-20°C
Actn3
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.