missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha Actinin 3 Recombinant Rabbit Monoclonal Antibody (23GB3625), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552587
This item is not returnable.
View return policy
Description
Alpha-actinin-3, also known as alpha-actinin skeletal muscle isoform 3 or F-actin cross-linking protein, is a protein that in humans is encoded by the ACTN3 gene. This gene encodes a member of the alpha-actin binding protein gene family. The encoded protein is primarily expressed in skeletal muscle and functions as a structural component of sarcomeric Z line. This protein is involved in crosslinking actin containing thin filaments. An allelic polymorphism in this gene results in both coding and non-coding variants; the reference genome represents the coding allele.
Specifications
| alpha Actinin 3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Actn3 | |
| actinin alpha 3; actinin alpha 3 (gene/pseudogene); Actinin alpha3; actinin, alpha 3; Actinin-alpha3; ACTN3; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 3; alpha-Actinin3; Alpha-actinin-3; alphaActinin-3; F-actin cross-linking protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 171009, 89 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 23GB3625 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q08043 | |
| Actn3 | |
| A synthetic peptide of human ACTN3 (760-800AA). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction