missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha 1B-Glycoprotein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57969
This item is not returnable.
View return policy
Description
alpha 1B-Glycoprotein Polyclonal specifically detects alpha 1B-Glycoprotein in Human samples. It is validated for Western Blot.
Specifications
| A1BG | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| A1B, ABG, alpha-1-B glycoproteinGAB, alpha-1B-glycoprotein, DKFZp686F0970, HYST2477 | |
| Rabbit | |
| 54 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%. | |
| Human | |
| Purified |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P04217 | |
| A1BG | |
| Synthetic peptides corresponding to A1BG(alpha-1-B glycoprotein) The peptide sequence was selected from the N terminal of A1BG (NP_570602). Peptide sequence ITPGLKTTAVCRGVLRGVTFLLRREGDHEFLEVPEAQEDVEATFPVHQPG. | |
| Protein A purified | |
| RUO | |
| 1 | |
| Centrifuge vial prior to reconstitution. Reconstitute in 100μL to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction