missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-1A Adrenergic R/ADRA1A Antibody (S04-8B8), Novus Biologicals™
Rabbit Monoclonal Antibody
£345.00 - £528.00
Specifications
| Antigen | alpha-1A Adrenergic R/ADRA1A |
|---|---|
| Clone | S04-8B8 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615364
|
Novus Biologicals
NBP3-19566-100ul |
100 μg |
£528.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631595
|
Novus Biologicals
NBP3-19566-50ul |
50 μg |
£345.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
alpha-1A Adrenergic R/ADRA1A Monoclonal antibody specifically detects alpha-1A Adrenergic R/ADRA1A in Human, Mouse, Rat samples. It is validated for Western BlotSpecifica
| alpha-1A Adrenergic R/ADRA1A | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ADRA1Cadrenergic, alpha-1A-, receptor variant 1, ADRA1L1, adrenergic, alpha -1A-, receptor, adrenergic, alpha-1A-, receptor, adrenergic, alpha-1A-, receptor variant 3, adrenergic, alpha-1A-, receptor variant 5, adrenergic, alpha-1A-, receptor variant 8, adrenergic, alpha-1C-, receptor, alpha-1A adrenergic receptor, Alpha-1A adrenoceptor, Alpha-1A adrenoreceptor, ALPHA1AAR, Alpha-1C adrenergic receptor, Alpha-adrenergic receptor 1c, G protein coupled receptor | |
| A synthetic peptide of human alpha-1A Adrenergic R/ADRA1A (Uniprot # P35348) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S04-8B8 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, GPCR, Neuroscience | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 148 | |
| IgG | |
| Affinity purified |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto