missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-1A Adrenergic R/ADRA1A Antibody (S04-8B8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19566-50ul
This item is not returnable.
View return policy
Description
alpha-1A Adrenergic R/ADRA1A Monoclonal antibody specifically detects alpha-1A Adrenergic R/ADRA1A in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| alpha-1A Adrenergic R/ADRA1A | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S04-8B8 | |
| Unconjugated | |
| ADRA1Cadrenergic, alpha-1A-, receptor variant 1, ADRA1L1, adrenergic, alpha -1A-, receptor, adrenergic, alpha-1A-, receptor, adrenergic, alpha-1A-, receptor variant 3, adrenergic, alpha-1A-, receptor variant 5, adrenergic, alpha-1A-, receptor variant 8, adrenergic, alpha-1C-, receptor, alpha-1A adrenergic receptor, Alpha-1A adrenoceptor, Alpha-1A adrenoreceptor, ALPHA1AAR, Alpha-1C adrenergic receptor, Alpha-adrenergic receptor 1c, G protein coupled receptor | |
| A synthetic peptide of human alpha-1A Adrenergic R/ADRA1A (Uniprot # P35348) | |
| 50 μg | |
| Apoptosis, GPCR, Neuroscience | |
| 148 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction