missing translation for 'onlineSavingsMsg'
Learn More

Alpha-(1-Hydroxycyclohexyl)-4-Methoxy-N,N-Dimethylbenzeneacetamide, TRC

Product Code. 30392658
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392658 50 mg
30389891 10 mg
30398835 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392658 Supplier Toronto Research Chemicals Supplier No. TRCH55555550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 295366-48-2
Molecular Formula C17H25NO3
Molecular Weight (g/mol) 291.39
Synonym alpha-(1-Hydroxycyclohexyl)-4-Methoxy-N,N-Dimethylbenzeneacetamide
IUPAC Name 2-(1-hydroxycyclohexyl)-2-(4-methoxyphenyl)-N,N-dimethylacetamide
SMILES COc1ccc(cc1)C(C(=O)N(C)C)C2(O)CCCCC2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H25NO3/c1-18(2)16(19)15(17(20)11-5-4-6-12-17)13-7-9-14(21-3)10-8-13/h7-10,15,20H,4-6,11-12H2,1-3H3
Formula Weight 291.1834
Chemical Name or Material alpha-(1-Hydroxycyclohexyl)-4-Methoxy-N,N-Dimethylbenzeneacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.