CAS RN 295366-48-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Alpha-(1-Hydroxycyclohexyl)-4-Methoxy-N,N-Dimethylbenzeneacetamide, TRC
CAS: 295366-48-2 Molecular Formula: C17H25NO3 Molecular Weight (g/mol): 291.39 Synonym: alpha-(1-Hydroxycyclohexyl)-4-Methoxy-N,N-Dimethylbenzeneacetamide,alpha-(1-Hydroxycyclohexyl)-4-Methoxy-N,N-Dimethylbenzeneacetamide IUPAC Name: 2-(1-hydroxycyclohexyl)-2-(4-methoxyphenyl)-N,N-dimethylacetamide SMILES: COc1ccc(cc1)C(C(=O)N(C)C)C2(O)CCCCC2