missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha 1 Antitrypsin Rabbit anti-Human, Mouse, Rat, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | Alpha 1 Antitrypsin |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | CoraLite 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17741269
|
Proteintech
CL488-16382-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17731269
|
Proteintech
CL488-16382-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alpha 1 Antitrypsin Polyclonal antibody specifically detects Alpha 1 Antitrypsin in Human, Mouse, Rat samples. It is validated for FC (Intra)Specifications
| Alpha 1 Antitrypsin | |
| Flow Cytometry | |
| CoraLite 488 | |
| Rabbit | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| IgG | |
| Antigen affinity purification | |
| Antibody |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| 5265 | |
| Primary | |
| Store at -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title