missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha 1 Antitrypsin Rabbit anti-Human, Mouse, Rat, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-16382-20UL
This item is not returnable.
View return policy
Description
Alpha 1 Antitrypsin Polyclonal antibody specifically detects Alpha 1 Antitrypsin in Human, Mouse, Rat samples. It is validated for FC (Intra)Specifications
| Alpha 1 Antitrypsin | |
| Polyclonal | |
| CoraLite 488 | |
| BC000484 | |
| Antigen affinity purification | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| Rabbit | |
| 20 μL | |
| 5265 | |
| Store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction