missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha 1 Antichymotrypsin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12192-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a plasma protease inhibitor and member of the serine protease inhibitor class. Polymorphisms in this protein appear to be tissue specific and influence protease targeting. Variations in this proteiNULLs sequence have been implicated in Alzheimer′s disease, and deficiency of this protein has been associated with liver disease. Mutations have been identified in patients with Parkinson disease and chronic obstructive pulmonary disease.Specifications
| alpha 1 Antichymotrypsin | |
| Polyclonal | |
| Unconjugated | |
| SERPINA3 | |
| A1 Antichymotrypsin, AACT, ACT, Alpha 1 antichymotrypsin, GIG24, GIG25, Serpin A3, SERPINA3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01011 | |
| SERPINA3 | |
| alpha 1 Antichymotrypsin Fusion Protein Ag2830 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction