missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha 1 Antichymotrypsin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | alpha 1 Antichymotrypsin |
|---|---|
| Concentration | 0.47 mg/mL |
| Applications | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16859753
|
Proteintech
12192-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16849753
|
Proteintech
12192-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a plasma protease inhibitor and member of the serine protease inhibitor class. Polymorphisms in this protein appear to be tissue specific and influence protease targeting. Variations in this proteiNULLs sequence have been implicated in Alzheimer′s disease, and deficiency of this protein has been associated with liver disease. Mutations have been identified in patients with Parkinson disease and chronic obstructive pulmonary disease.Specifications
| alpha 1 Antichymotrypsin | |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P01011 | |
| 12 | |
| alpha 1 Antichymotrypsin Fusion Protein Ag2830 | |
| Primary | |
| -20°C | |
| SERPINA3 |
| 0.47 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A1 Antichymotrypsin, AACT, ACT, Alpha 1 antichymotrypsin, GIG24, GIG25, Serpin A3, SERPINA3 | |
| SERPINA3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel