Learn More
Invitrogen™ ALP Monoclonal Antibody (K52014_9F2), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | ALP |
|---|---|
| Clone | K52014_9F2 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30203360
|
Invitrogen™
MA555956 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30200697
|
Invitrogen™
MA556336 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: MNPIVVVHGG GAGPISKDRK ERVHQGMVRA ATVGYGILRE GGSAVDAVEG AVVALEDDPE FNAGCGSVLN TNGEVEMDAS IMDGKDLSAG AVSAVQCIAN PIKLARLVME KTPHCFLTDQ GAAQFAAAMG VPEIPGEKLV TERNKKRLEK EKHEKGAQKT DCQKNLGTVG AVALDCKGNV AYATSTGGIV NKMVGRVGDS PCLGAGGYAD NDIGAVSTTG HGESILKVNL ARLTLFHIEQ GKTVEEAADL SLGYMKSRVK GLGGLIVVSK TGDWVAKWTS TSMPWAAAKD GKLHFGIDPD DTTITDLP
ALP Monoclonal antibody specifically detects ALP in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ALP | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 2410004D18Rik; ALP; ALP1; asparaginase and isoaspartyl peptidase 1; asparaginase like 1; asparaginase-like 1; asparaginase-like 1 protein; asparaginase-like protein 1; Asparaginase-like sperm autoantigen; ASRGL1; AU040643; AW060375; Beta-aspartyl-peptidase; CRASH; FLJ22316; Glial asparaginase; Gliap; Hiob; Hiob2; isoaspartyl dipeptidase; Isoaspartyl peptidase/L-asparaginase; Isoaspartyl peptidase/L-asparaginase alpha chain; Isoaspartyl peptidase/L-asparaginase beta chain; L-asparaginase; L-asparagine amidohydrolase; testis secretory sperm-binding protein Li 242mP; zgc:103568 | |
| Recombinant human ASRGL1 (immunogen range is 1-308). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Asrgl1 |
| K52014_9F2 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q7L266 | |
| 80150 | |
| IgG2a | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.