Learn More
Invitrogen™ ALP Monoclonal Antibody (K52014_9F2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556336
Description
Sequence of this protein is as follows: MNPIVVVHGG GAGPISKDRK ERVHQGMVRA ATVGYGILRE GGSAVDAVEG AVVALEDDPE FNAGCGSVLN TNGEVEMDAS IMDGKDLSAG AVSAVQCIAN PIKLARLVME KTPHCFLTDQ GAAQFAAAMG VPEIPGEKLV TERNKKRLEK EKHEKGAQKT DCQKNLGTVG AVALDCKGNV AYATSTGGIV NKMVGRVGDS PCLGAGGYAD NDIGAVSTTG HGESILKVNL ARLTLFHIEQ GKTVEEAADL SLGYMKSRVK GLGGLIVVSK TGDWVAKWTS TSMPWAAAKD GKLHFGIDPD DTTITDLP
ALP is found in a variety of tumors but only occasionally in normal tissues. Ovarian cancers are highly positive. ALP is further found in brain tumors, some colonic cancers, some lung cancers, some prostate cancers, uterine cancers, some breast cancers and some thyroid cancers. It was not found in serum.
Specifications
| ALP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q7L266 | |
| Asrgl1 | |
| Recombinant human ASRGL1 (immunogen range is 1-308). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| K52014_9F2 | |
| Unconjugated | |
| Asrgl1 | |
| 2410004D18Rik; ALP; ALP1; asparaginase and isoaspartyl peptidase 1; asparaginase like 1; asparaginase-like 1; asparaginase-like 1 protein; asparaginase-like protein 1; Asparaginase-like sperm autoantigen; ASRGL1; AU040643; AW060375; Beta-aspartyl-peptidase; CRASH; FLJ22316; Glial asparaginase; Gliap; Hiob; Hiob2; isoaspartyl dipeptidase; Isoaspartyl peptidase/L-asparaginase; Isoaspartyl peptidase/L-asparaginase alpha chain; Isoaspartyl peptidase/L-asparaginase beta chain; L-asparaginase; L-asparagine amidohydrolase; testis secretory sperm-binding protein Li 242mP; zgc:103568 | |
| Mouse | |
| Protein A | |
| RUO | |
| 80150 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.