missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALKBH3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57586
This item is not returnable.
View return policy
Description
ALKBH3 Polyclonal specifically detects ALKBH3 in Human samples. It is validated for Western Blot.
Specifications
| ALKBH3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ABH3MGC118793, alkB, alkylation repair homolog 3 (E. coli), Alkylated DNA repair protein alkB homolog 3, DEPC1alpha-ketoglutarate-dependent dioxygenase alkB homolog 3, DEPC-1MGC118792, EC 1.14.11, EC 1.14.11.-, FLJ43614, MGC118790, PCA1, Prostate cancer antigen 1, prostate cancer antigen-1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Equine: 100%; Human: 100%; Mouse: 100%; Xenopus: 92%; Chicken: 92%; Canine: 92%; Guinea pig: 92%; Rabbit: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96Q83 | |
| ALKBH3 | |
| Synthetic peptides corresponding to ALKBH3(alkB, alkylation repair homolog 3 (E. coli)) The peptide sequence was selected from the middle region of ALKBH3. Peptide sequence EMRKKPPPEENGDYTYVERVKIPLDHGTLLIMEGATQADWQHRVPKEYHS. | |
| 100 μL | |
| Proteases & Other Enzymes | |
| 221120 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction