missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Alkaline Peptone Water (Bottled)

Product Code. 10699944
This item is not returnable. View return policy
Product Code. 10699944 Supplier Thermo Scientific™ Supplier No. BO0335E

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Enrich and recover Vibrio species from food, water and clinical samples using these ready prepared bottles.

Enrich and recover Vibrio species from food, water and clinical samples with Thermo Scientific™ Oxoid™ Alkaline Peptone Water ready prepared bottles. The medium is selective by exploiting the ability of Vibrio species to tolerate alkaline conditions and high sodium chloride levels. The peptone content provides the conditions for growth and recovery of Vibrio species, supplying vitamins, minerals, and nitrogen in the form of amino acids.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready prepared bottles of Alkaline Peptone Water.

  • Creates strong alkaline and high sodium conditions to provide a selective environment for Vibrio
  • Can be used for direct microscopic examination of samples using the hanging drop method
  • Convenient: the pre-filled bottles are ready for immediate use
  • Simple: easy-to-use with minimum training
  • Long shelf life: up to 12 months

Alkaline Peptone Water was first formulated by Shread, Donovan and Lee1 to be used as a non-selective enrichment broth for the cultivation of Aeromonas spp. Cruickshank reported that when the pH is raised, the medium can be used to effectively cultivate Vibrio species.2 The 2% (w/v) sodium chloride incorporated in this medium promotes the growth of Vibrio cholerae, while the alkalinity of this medium inhibits most of the unwanted background flora.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Color Straw, clear
Content And Storage 2 to 25°C
Description Alkaline Peptone Water (Bottled)
Format Universal bottles
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 10.0g + 0.3g
Packaging Type Twenty-four universal bottles with screw cap closures in a box.
Quantity 24/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.