missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALK/CD246 Antibody (5A4), Novus Biologicals™
Mouse Monoclonal Antibody
£156.00 - £718.00
Specifications
| Antigen | ALK/CD246 |
|---|---|
| Clone | 5A4 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:25 - 1:70 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18284751
|
Novus Biologicals
NB120-17127-0.1ML |
0.1 mL |
£156.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18263673
|
Novus Biologicals
NB120-17127-0.25ML |
0.25 mL |
£388.00
0.25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18537552
|
Novus Biologicals
NB120-17127 |
0.5 mL |
£718.00
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALK/CD246 Monoclonal specifically detects ALK/CD246 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ALK/CD246 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:25 - 1:70 | |
| Monoclonal | |
| Supernatant | |
| Human | |
| ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3 | |
| ALK | |
| IgG1 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 5A4 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Tissue culture supernatant with 0.09% sodium azide | |
| 238 | |
| Prokaryotic recombinant protein corresponding to a region which spans the tyrosine kinase catalytic domain and part of the C-terminus, corresponding to amino acids 419-520 of Human ALK. | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title