missing translation for 'onlineSavingsMsg'
Learn More

ALK/CD246 Antibody (5A4), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NB120-17127

 View more versions of this product

Product Code. 18537552

  • £718.00 / 0.50mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ALK/CD246 Monoclonal specifically detects ALK/CD246 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ALK/CD246
Monoclonal
Unconjugated
Tissue culture supernatant with 0.09% sodium azide
ALK
Prokaryotic recombinant protein corresponding to a region which spans the tyrosine kinase catalytic domain and part of the C-terminus, corresponding to amino acids 419-520 of Human ALK.
Primary
Human
Supernatant
Immunohistochemistry, Immunohistochemistry (Paraffin)
5A4
Immunohistochemistry, Immunohistochemistry-Paraffin 1:25 - 1:70
ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3
Mouse
0.5 mL
238
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only