Learn More
Agilent BioTek Alignment Tool for Imaging Multi-Mode Reader
GREENER_CHOICE
Product Code. 16522132 Shop All Agilent BioTek Products
Click to view available options
Product Type:
Alignment Tool for 10X/20X Phase Contrast Objectives
Alignment Tool for 40X Phase Contrast Objective
Unit Size:
Each
Product Code. 16522132
Brand: Agilent BioTek 1322507
For use with Cytation 5 Cell Imaging Multi-Mode Readers.
For use with Cytation 5 Cell Imaging Multi-Mode Readers.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Alignment Tool for 40X Phase Contrast Objective
For Use With (Equipment) Cytation 5 Cell Imaging Multi-Mode Reader