missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALG13 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92394-0.1ml
This item is not returnable.
View return policy
Description
ALG13 Polyclonal antibody specifically detects ALG13 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ALG13 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Asparagine-linked glycosylation 13 homolog, asparagine-linked glycosylation 13 homolog (S. cerevisiae), chromosome X open reading frame 45, CXorf45, EC 2.4.1.141, FLJ23018, FLJ31785, GLT28D1, glycosyltransferase 28 domain containing 1, Glycosyltransferase 28 domain-containing protein 1, hematopoietic stem/progenitor cells protein MDS031, MDS031, MGC12423, UDP-N-acetylglucosamine transferase subunit ALG13 homolog, YGL047W | |
| A synthetic peptide corresponding to a sequence within amino acids 100-167 of human ALG13 (NP_001311219.1). VVINEKLMNNHQLELAKQLHKEGHLFYCTCSTLPGLLQSMDLSTLKCYPPGQPEKFSAFLDKVVGLQK | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79868 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction