Learn More
Thermo Scientific™ Yttria Stabilized Zirconium Oxide Conical Crucibles, High Form
Brand:  Thermo Scientific™ 041289.KF
 View more versions of this product
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15453835
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Zirconium Oxide
50 mL
58mm
34mm
1 Piece
51mm
Yttria Stabilized Zirconium Oxide Conical Crucibles, High Form