missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 532 NHS Ester (Succinimidyl Ester)

Product Code. 11574007
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
500mL
5mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11574007

Brand: Invitrogen™ A20101MP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Alexa Fluor™ 532 is a bright yellow dye with excitation ideally suited for the frequency-doubled Nd:YAG laser line.

Alexa Fluor™ 532 is a bright yellow dye with excitation ideally suited for the frequency-doubled Nd:YAG laser line. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 532 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 532 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection.

The NHS ester (or succinimidyl ester) of Alexa Fluor™ 532 is the most popular tool for conjugating this dye to a protein or antibody. NHS esters can be used to label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugate will exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ NHS ester:

  • Fluorophore label: Alexa Fluor™ 532 dye
  • Reactive group: NHS ester
  • Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
  • Ex/Em of the conjugate: 530/555 nm
  • Extinction coefficient: 81,000 cm-1M-1
  • Molecular weight: 723.8

Typical Conjugation Reaction

You can conjugate amine-reactive reagents with virtually any protein or peptide (the provided protocol is optimized for IgG antibodies). You can scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) (D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification

Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:

  • Antibody Conjugate Purification Kit for 0.5-1 mg (Cat. No. A33086)
  • Antibody Conjugate Purification Kit for 20-50 μg (A33087)
  • Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling

We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We'll Make a Custom Conjugate for You

If you can't find what you're looking for in our online catalog, we'll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type Alexa Fluor™ 532 NHS Ester (Succinimidyl Ester)
Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Alexa Fluor™ 532
Label Type Alexa Fluor
Product Line Alexa Fluor
Emission 555 nm
Excitation 530 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.