missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 488 Hydroxylamine

Product Code. 10226602
Click to view available options
Quantity:
1 mg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10226602

Brand: Invitrogen™ A30629

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Alexa Fluor™ 488 Hydroxylamine is useful as a cell tracer and as a reactive dye for labeling aldehydes or ketones in polysaccharides or glycoproteins.

Alexa Fluor™ 488 Hydroxylamine is useful as a cell tracer and as a reactive dye for labeling aldehydes or ketones in polysaccharides or glycoproteins. Alexa Fluor™ 488 is a bright, green fluorescent dye with excitation ideally suited to the 488 nn laser line. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 488 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 488 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection.

Detailed information about this AlexaFluor™ Hydroxylamine:

  • Fluorophore label: Alexa Fluor™ 488 dye
  • Reactive group: Hydroxylamine
  • Reactivity: Aldehydes or ketones
  • Ex/Em of the conjugate: 494/518 nm
  • Extinction coefficient: 77,000 cm-1M-1
  • Spectrally similar dyes: FITC, GFP
  • Molecular weight: 895.07

Cell Tracking and Tracing Applications

Alexa Fluor™ hydrazides and Hydroxylamines are useful as low molecular weight, membrane-impermeant, aldehyde-fixable cell tracers, exhibiting brighter fluorescence and greater photostability than cell tracers derived from other spectrally similar fluorophores. They are easily loaded into cells by microinjection, infusion from patch pipette, or uptake induced by our Influx™ Pinocytic Cell-Loading Reagent (Cat. No. I14402).

Glycoprotein and Polysaccharide Labeling Applications

The Alexa Fluor™ hydrazides and Hydroxylamines are reactive molecules that can be used to add a fluorescent label to biomolecules containing aldehydes or ketones. Aldehydes and ketones can be introduced into polysaccharides and glycoproteins by periodate-mediated oxidation of vicinal diols. Galactose oxidase can also be used to oxidize terminal galactose residues of glycoproteins to aldehydes.

Hydrazide vs Hydroxylamine

Hydrazine derivatives react with ketones and aldehydes to yield relatively stable hydrazones. Hydroxylamine derivatives (aminooxy compounds) react with aldehydes and ketones to yield oximes. Oximes are superior to hydrazones with respect to hydrolytic stability. Both hydrazones and oximes can be reduced with sodium borohydride (NaBH4) to further increase the stability of the linkage.

Learn More About Protein and Antibody Labeling

We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We'll Make a Custom Conjugate for You

If you can't find what you're looking for in our online catalog, we'll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

Related Products

  • DMSO (dimethylsulfoxide) (Cat. No. D12345)
  • Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
  • Antibody Conjugate Purification Kit for 20-50 μg (A33087)
  • Antibody Conjugate Purification kit for 50-100 μg (A33088)
TRUSTED_SUSTAINABILITY

Specifications

Description Alexa Fluor™ 488 Hydroxylamine, 1mg, Yellow, Solid, C27H38N4O8S Molecular Formula, 578.68 Molecular Weight, Alexa Fluor™ 488 Label, Aldehyde, Carboxylic Acid, Ketone Chemical Reactivity, 494/518nm Excitation/Emission Wavelength, Used for Label
Formulation Solid
For Use With (Application) Cell Viability, Proliferation and Function, Labeling Chemistry, Protein Labeling and Crosslinking
Quantity 1 mg
Product Type Hydroxylamine
pH Range 4 to 10
Molecular Weight (g/mol) 578.68kDa
Purity HPLC, 99% at 254nm
Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Shipping Condition Room Temperature
Reactive Moiety Amine, Hydroxylamine
Label or Dye Alexa Fluor™ 488
Label Type Alexa Fluor
Color Yellow
Product Line Alexa Fluor
Emission 518
Excitation 494 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.