missing translation for 'onlineSavingsMsg'
Learn More

Aldo-keto Reductase 1C1/AKR1C1 Antibody (AKR1C1/9314), Novus Biologicals™

Mouse Monoclonal Antibody

£195.00 - £368.00

Specifications

Antigen Aldo-keto Reductase 1C1/AKR1C1
Clone AKR1C1/9314
Concentration 0.2 mg/ml
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30298760
View Documents
Novus Biologicals
NBP3-44592-20ug
20 μg
£195.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30293711
View Documents
Novus Biologicals
NBP3-44592-100ug
100 μg
£368.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aldo-keto Reductase 1C1/AKR1C1 Monoclonal antibody specifically detects Aldo-keto Reductase 1C1/AKR1C1 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

Aldo-keto Reductase 1C1/AKR1C1
0.2 mg/ml
Monoclonal
Purified
RUO
Human
20 alpha-hydroxysteroid dehydrogenase, 20-ALPHA-HSD, 20-alpha-hydroxysteroid dehydrogenase, aldo-keto reductase family 1 member C1, aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; 20-alpha(3-alpha)-hydroxysteroid dehydrogenase), C9, Chlordecone reductase homolog HAKRC, DD1/DD2, DD1MGC8954, DDHH-37, dihydrodiol dehydrogenase 1, Dihydrodiol dehydrogenase 1/2, dihydrodiol dehydrogenase isoform DD1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.112, EC 1.1.1.149,2-ALPHA-HSD, EC 1.3.1.20, HAKRCDDH1aldo-keto reductase C, HBAB, hepatic dihydrodiol dehydrogenase, High-affinity hepatic bile acid-binding protein, Indanol dehydrogenase, MBAB, Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase, type II 3-alpha-hydroxysteroid dehydrogenase
Recombinant human full-length Aldo-keto Reductase 1C1/AKR1C1 protein (Uniprot: Q04828). Exact sequence is proprietary.
Primary
Store at 4⪚C.
AKR1C1/9314
Immunohistochemistry
Unconjugated
Mouse
metabolism
10 mM PBS with 0.05% BSA
1645
IgG2b κ
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.