missing translation for 'onlineSavingsMsg'
Learn More

Aldo-keto Reductase 1C1/AKR1C1 Antibody (AKR1C1/9314), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-44592-100ug

 View more versions of this product

Product Code. 30293711

  • £368.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Aldo-keto Reductase 1C1/AKR1C1 Monoclonal antibody specifically detects Aldo-keto Reductase 1C1/AKR1C1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aldo-keto Reductase 1C1/AKR1C1
Monoclonal
0.2 mg/ml
10 mM PBS with 0.05% BSA
Mouse
Protein A purified
RUO
Primary
Human
Purified
Immunohistochemistry
AKR1C1/9314
Unconjugated
20 alpha-hydroxysteroid dehydrogenase, 20-ALPHA-HSD, 20-alpha-hydroxysteroid dehydrogenase, aldo-keto reductase family 1 member C1, aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; 20-alpha(3-alpha)-hydroxysteroid dehydrogenase), C9, Chlordecone reductase homolog HAKRC, DD1/DD2, DD1MGC8954, DDHH-37, dihydrodiol dehydrogenase 1, Dihydrodiol dehydrogenase 1/2, dihydrodiol dehydrogenase isoform DD1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.112, EC 1.1.1.149,2-ALPHA-HSD, EC 1.3.1.20, HAKRCDDH1aldo-keto reductase C, HBAB, hepatic dihydrodiol dehydrogenase, High-affinity hepatic bile acid-binding protein, Indanol dehydrogenase, MBAB, Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase, type II 3-alpha-hydroxysteroid dehydrogenase
Recombinant human full-length Aldo-keto Reductase 1C1/AKR1C1 protein (Uniprot: Q04828). Exact sequence is proprietary.
100 μg
metabolism
1645
Store at 4⪚C.
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.